missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic, Puriss., meets analytical spec. of BP, Ph. Eur., USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance), Honeywell Fluka™

Catalog Number 15690750
Click to view available options
Quantity:
1 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
This item is not returnable. View return policy
7558-79-4
HNa2O4P
141.96
MFCD00003496
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
24203
CHEBI:34683
disodium;hydrogen phosphate
[Na+].[Na+].OP([O-])([O-])=O
This item is not returnable. View return policy
7558-79-4
HNa2O4P
141.96
MFCD00003496
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
24203
CHEBI:34683
disodium;hydrogen phosphate
[Na+].[Na+].OP([O-])([O-])=O

Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 4 x 5 kg
Linear Formula Na2HPO4
Solubility Information Very Soluble in hot water, 8 parts soluble in water , insoluble in alcohol
Formula Weight 141.96
Percent Purity 98 to 100.5% (calc. to the dried substance)
Grade puriss.
Chemical Name or Material Sodium Phosphate Dibasic, Meets analytical specification of Ph.Eur.BP, USP, FCC
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'