missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic, Puriss., meets analytical spec. of BP, Ph. Eur., USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance), Honeywell Fluka™
BIOPHARMA

Product Code. 15600760 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
Unit Size:
1kg
25kg
5kg
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15600760

Brand: Honeywell-Fluka 042765KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
IUPAC Name disodium;hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Physical Form Anhydrous
Quantity 5 kg
Linear Formula Na2HPO4
Solubility Information Very Soluble in hot water, 8 parts soluble in water , insoluble in alcohol
Formula Weight 141.96g/mol
Percent Purity 98 to 100.5% (calc. to the dried substance)
Grade puriss.
Chemical Name or Material Sodium Phosphate Dibasic, Meets analytical specification of Ph.Eur.BP, USP, FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.