missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Dibasic Heptahydrate (Colorless-to-White Crystals), Fisher BioReagents™
Description
Sodium Phosphate Dibasic Heptahydrate (Colorless-to-White Crystals)
This buffer component is commonly used in tissue culture and molecular biology applications.
Specifications
Specifications
| Melting Point | 48°C |
| Color | White |
| pH | 9.0 to 9.3 |
| Physical Form | Solid |
| Quantity | 500 g |
| Assay Percent Range | 98.0 to 102.0 % |
| Merck Index | 15, 8789 |
| Formula Weight | 268.07 |
| Percent Purity | 98.0 to 102.0% |
| Identification | Pass Test |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?