missing translation for 'onlineSavingsMsg'
Learn More

Melford Sodium Phosphate Dibasic Heptahydrate

Product Code. 30018790
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018790 5 kg 5kg
30018789 500 g 500g
30018788 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018790 Supplier Melford Supplier No. S231755000.0
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Sodium Phosphate Dibasic Heptahydrate, also known as disodium hydrogen phosphate heptahydrate, is a chemical compound with significant applications in the life science marketplace.

It is a hydrated form of sodium phosphate dibasic, meaning it contains seven water molecules per molecule of the compound.

Sodium Phosphate Dibasic Heptahydrate is primarily used as a buffering agent and as a source of phosphate ions. Buffers help maintain a stable pH in biological and biochemical processes, ensuring optimal conditions for enzymatic reactions, cell culture, and other molecular biology techniques.

It is also commonly used in laboratory settings for the preparation of buffer solutions with specific pH ranges and buffering capacities. It is particularly useful in applications where a higher pH range is required, as it has a buffering capacity in the alkaline pH range

Moreover, the compound is used in various biological and biochemical assays, protein purification processes, and the formulation of diagnostic kits and pharmaceutical products. It serves as a source of phosphate ions, which are essential for numerous biological processes such as DNA and RNA synthesis, energy metabolism, and signal transduction.

Specifications

Chemical Name or Material Sodium Phosphate Dibasic Heptahydrate
CAS 7782-85-6
Quantity 5 kg
Molecular Formula Na2HPO4-7H2O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.