missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic Dihydrate, TRC

Product Code. 30137460
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137460 5 g 5g
17994445 1 g 1g
17984715 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137460 Supplier Toronto Research Chemicals Supplier No. TRCS2245005G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10028-24-7
Molecular Formula Na2HPO4 . 2 (H2O)
Molecular Weight (g/mol) 177.99
Synonym Disodium Hydrogen Phosphate Dihydrate, Disodium Monohydrogen Phosphate Dihydrate, Disodium Orthophosphate Dihydrate, Disodium Phosphate Dihydrate, Sodium Phosphate (Na2HPO4) Dihydrate;
IUPAC Name disodium;hydrogen phosphate;dihydrate
SMILES O.O.[Na+].[Na+].OP(=O)([O-])[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/2Na.H3O4P.2H2O/c;;1-5(2,3)4;;/h;;(H3,1,2,3,4);2*1H2/q2*+1;;;/p-2
Formula Weight 177.9619
Chemical Name or Material Sodium Phosphate Dibasic Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.