missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic dihydrate, meets analytical spec. of BP, Ph. Eur., 98.5-101% (calc. to the dried substance), Honeywell Fluka™
BIOPHARMA

Product Code. 15680740 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
25kg
5kg
Pack of 4
Pack of 6
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15680740 4 x 5 kg Pack of 4
15650740 1 kg 1kg
15660740 2.5 kg 2.5kg
15690740 5 kg 5kg
15600750 6 x 1 kg Pack of 6
15610750 6 x 2.5 kg Pack of 6
15670740 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15680740 Supplier Honeywell-Fluka Supplier No. 042724X5KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Meets analytical specification of BP, Ph. Eur., 98.5-101% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
PubChem CID 6451167
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Quantity 4 x 5 kg
Linear Formula Na2HPO4 · 2H2O
IUPAC Name disodium;hydrogen phosphate;dihydrate
Formula Weight 177.99
Percent Purity 98.5 to 101% (calc. to the dried substance)
Chemical Name or Material Sodium Phosphate Dibasic Dihydrate, Meets analytical specification of Ph.Eur.BP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.