Learn More
Melford Sodium Phosphate Dibasic, Anhydrous
Description
Sodium Phosphate Dibasic Anhydrous is a chemical compound that is commonly known as disodium hydrogen phosphate or disodium phosphate.
This compound is an anhydrous form of sodium phosphate, meaning it does not contain any water molecules.
In the life science field, Sodium Phosphate Dibasic Anhydrous is primarily used as a buffering agent, which helps maintain a stable pH in various biological and biochemical processes.
It is often utilized in laboratory settings for the preparation of various biological reagents, cell culture media, and protein purification processes. The compound's buffering properties make it valuable in the formulation of diagnostic kits and pharmaceutical products, where maintaining a specific pH is critical for the stability and effectiveness of the final product
Spécification
Spécification
| Chemical Name or Material | Sodium Phosphate Dibasic, Anhydrous |
| CAS | 7558-79-4 |
| Quantity | 500 g |
| Molecular Formula | Na2HPO4 |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.