missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, dibasic, 99+%, for analysis, anhydrous

Product Code. 10698504 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Plastic Bottle
Unit Size:
100g
500g
5kg
This item is not returnable. View return policy

Product Code. 10698504

Brand: Thermo Scientific Acros 204850050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7558-79-4 | HNa2O4P | 141.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Color White
pH 8.7 to 9.3 (5% soln. at 20°C)
Physical Form Granular Powder
Quantity 5 kg
Assay Percent Range 99+%
Linear Formula Na2HPO4
Merck Index 15, 8789
Solubility Information Solubility in water: 75g/L (20°C). Other solubilities: practically insoluble in alcohol
IUPAC Name disodium;hydrogen phosphate
Formula Weight 141.96
Percent Purity ≥99%
Grade Analytical
Loss on Drying 0.2% max.
Packaging Plastic Bottle
Chemical Name or Material Sodium phosphate, dibasic, p.a.
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.