missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate ACS AR, Macron Fine Chemicals™

Product Code. 15538454 Shop All J.T.Baker Products
Change view
Click to view available options
POLYSTORMOR; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15538454 POLYSTORMOR; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15538454 Supplier Avantor Macron Supplier No. 791404

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7782-85-6
Quantity 500 g
Molecular Formula H15Na2O11P
MDL Number MFCD00149180
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
SMILES O.[Na+].[Na+].OP([O-])([O-])=O
IUPAC Name disodium phosphoric acid heptahydrate
Molecular Weight (g/mol) 268.06
PubChem CID 6096963
Formula Weight 268.07
Grade ACS
Packaging POLYSTORMOR
Chemical Name or Material Sodium Phosphate, Dibasic, Heptahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.