missing translation for 'onlineSavingsMsg'
Learn More

Sodium phenoxide, TRC

Product Code. 30423493
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
1g
2.5g
Product Code. Quantity unitSize
30423493 1 g 1g
30424508 2.5 g 2.5g
2 options
This item is not returnable. View return policy

Product Code. 30423493

Brand: Toronto Research Chemicals TRCS2244351G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-02-6
Formula Weight 116.0238
InChI Formula InChI=1S/C6H6O.Na/c7-6-4-2-1-3-5-6;/h1-5,7H;/q;+1/p-1
IUPAC Name sodium;phenoxide
Molecular Formula C6H5ONa
Molecular Weight (g/mol) 116.09
SMILES [Na+].[O-]c1ccccc1
Synonym Phenol, sodium salt (1:1) (ACI), Phenol, sodium salt (8CI, 9CI), Sodium phenoxide (6CI), Phenol sodium, Sodium carbolate, Sodium phenate, Sodium phenolate, Sodium phenylate
Chemical Name or Material Sodium phenoxide

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.