missing translation for 'onlineSavingsMsg'
Learn More

Sodium Persulfate, TRC

Product Code. 30425804
Click to view available options
Quantity:
25 g
50 g
Unit Size:
25g
50g
This item is not returnable. View return policy

Product Code. 30425804

Brand: Toronto Research Chemicals TRCS66590025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Persulfate
CAS 7775-27-1
Molecular Formula Na2O8S2
Quantity 25 g
Synonym Peroxydisulfuric Acid ([(HO)S(O)2]2O2) Disodium Salt, Sodium Peroxydisulfate, Disodium Peroxodisulfate, Disodium Peroxydisulfate, Disodium Persulfate, SP, SP (Peracid), SPS, Sodium Dipersulfate, Sodium Peroxodisulfate, Sodium Peroxydisulfate (Na2S2O8), Sodium Persulfate (Na2S2O8)
InChI Formula InChI=1S/2Na.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2
SMILES [Na+].[Na+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
IUPAC Name disodium;sulfonatooxy sulfate
Molecular Weight (g/mol) 238.1
Formula Weight 237.883
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.