missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Persulfate, TRC
Click to view available options
Quantity:
25 g
50 g
Unit Size:
25g
50g
Specifications
Specifications
| Chemical Name or Material | Sodium Persulfate |
| CAS | 7775-27-1 |
| Molecular Formula | Na2O8S2 |
| Quantity | 50 g |
| Synonym | Peroxydisulfuric Acid ([(HO)S(O)2]2O2) Disodium Salt, Sodium Peroxydisulfate, Disodium Peroxodisulfate, Disodium Peroxydisulfate, Disodium Persulfate, SP, SP (Peracid), SPS, Sodium Dipersulfate, Sodium Peroxodisulfate, Sodium Peroxydisulfate (Na2S2O8), Sodium Persulfate (Na2S2O8) |
| InChI Formula | InChI=1S/2Na.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2 |
| SMILES | [Na+].[Na+].[O-]S(=O)(=O)OOS(=O)(=O)[O-] |
| IUPAC Name | disodium;sulfonatooxy sulfate |
| Molecular Weight (g/mol) | 238.1 |
| Formula Weight | 237.883 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction