missing translation for 'onlineSavingsMsg'
Learn More

Sodium Persulfate, TRC

Product Code. 30416484
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416484 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416484 Supplier Toronto Research Chemicals Supplier No. TRCS6659005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Persulfate
CAS 7775-27-1
Quantity 5 g
Molecular Formula Na2O8S2
Synonym Peroxydisulfuric Acid ([(HO)S(O)2]2O2) Disodium Salt, Sodium Peroxydisulfate, Disodium Peroxodisulfate, Disodium Peroxydisulfate, Disodium Persulfate, SP, SP (Peracid), SPS, Sodium Dipersulfate, Sodium Peroxodisulfate, Sodium Peroxydisulfate (Na2S2O8), Sodium Persulfate (Na2S2O8)
InChI Formula InChI=1S/2Na.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2
SMILES [Na+].[Na+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
IUPAC Name disodium;sulfonatooxy sulfate
Molecular Weight (g/mol) 238.1
Formula Weight 237.883
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.