missing translation for 'onlineSavingsMsg'
Learn More

Sodium perrhenate, 99.95% (metals basis)

Product Code. 11364769
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364769 1 g 1g
11354769 0.25 g 0.25g
11374769 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11364769 Supplier Thermo Scientific Alfa Aesar Supplier No. 062107.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13472-33-8 | O3Re | 234.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-33-8

Specifications

Melting Point 300°C
Color White
Physical Form Crystalline
Quantity 1 g
Molecular Formula NaO4Re
Linear Formula NaReO4
MDL Number MFCD00014231
UN Number UN1479
Synonym sodium perrhenate, unii-pe1t8nn47q, sodium tetraoxorhenate 1-, pe1t8nn47q, na.o4re, rhenium vii sodium oxide, rhenate reo41-, sodium, t-4, acmc-1bwm0, perrhenic acid sodium salt, sodium oxido trioxo rhenium
InChI Key KMISVFTVBOPTAI-UHFFFAOYSA-N
SMILES [O-][Re](=O)(=O)=O.[Na+]
IUPAC Name sodium;oxido(trioxo)rhenium
Molecular Weight (g/mol) 273.193
PubChem CID 5107658
Formula Weight 273.19
Percent Purity 99.95%
Odor Odorless
Sensitivity Hygroscopic
Density 5.390
Assay (metals basis)
Chemical Name or Material Sodium perrhenate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.