missing translation for 'onlineSavingsMsg'
Learn More

Sodium Peroxide, TRC

Product Code. 30413643
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413643 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413643 Supplier Toronto Research Chemicals Supplier No. TRCS66580025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium peroxide
CAS 1313-60-6
Quantity 25 g
Molecular Formula Na O2
Synonym Disodium Dioxide, Disodium Peroxide, Sodium Oxide (Na2O2)
InChI Formula InChI=1S/Na.O2/c;1-2/q+1;-2
SMILES [Na+].[O-][O-]
IUPAC Name sodium;peroxide
Molecular Weight (g/mol) 54.9891
Formula Weight 54.9801
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.