missing translation for 'onlineSavingsMsg'
Learn More

Sodium Peroxide, purum p.a, ≥95%, Solstice

Product Code. 15626860 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15626860 1 kg Poly bottle 1kg
15636860 250 g Poly bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15626860 Supplier Honeywell-Fluka Supplier No. 718831KG
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

for Wurzschmitt-decomposition, ACS reagent, beads (small),≥95%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-60-6
Molecular Formula Na2O2
Molecular Weight (g/mol) 77.978
MDL Number MFCD00003497
InChI Key PFUVRDFDKPNGAV-UHFFFAOYSA-N
Synonym sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
PubChem CID 14803
IUPAC Name disodium;peroxide
SMILES [O-][O-].[Na+].[Na+]

Specifications

Melting Point 300°C
Color White
Physical Form Crystals
Quantity 1 kg
UN Number UN1504
Solubility Information 106.7g/L at 20°C
Formula Weight 77.98g/mol
Percent Purity ≥95%
Packaging Poly bottle
Chemical Name or Material Sodium peroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.