missing translation for 'onlineSavingsMsg'
Learn More

Sodium Peroxide, Certified AR for Analysis, Fisher Chemical

Product Code. 10488030 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10488030 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10488030 Supplier Fisher Chemical Supplier No. S/6041/53
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

Melting Point 460°C
CAS 1313-60-6
Color Yellow
Physical Form Solid
Quantity 500 g
Molecular Formula Na2O2
MDL Number 3497
Synonym sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
InChI Key PFUVRDFDKPNGAV-UHFFFAOYSA-N
SMILES [O-][O-].[Na+].[Na+]
IUPAC Name disodium;peroxide
Molecular Weight (g/mol) 77.978
PubChem CID 14803
Formula Weight 77.97g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium peroxide
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 1
  • Skin corrosion/irritation Category 1 A
  • Substances and mixtures corrosive to metals Category 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.