missing translation for 'onlineSavingsMsg'
Learn More

Sodium peroxide, ACS, 93% min

Product Code. 11397708
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397708 10 g 10g
11387708 100 g 100g
11307718 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397708 Supplier Thermo Scientific Alfa Aesar Supplier No. 033384.09
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

For Use As An Oxidizing Agent, a Bleaching Agent, Deodorant | CAS: 1313-60-6 | Na2O2 | 77.978 g/mol

As an oxidizing agent, a bleaching agent, deodorant, pharmaceuticals and dyes, and as an analytical reagent.Sodium peroxide acts as a bleaching agent in laundry and wood pulp. It finds application in the extraction of minerals from various ores. It acts as an oxidizing agent as well as an oxygen source by reacting with carbon dioxide. Furthermore, it is used as a deodorant and an analytical reagent. It is also useful in scuba gear and in submarines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-60-6
Molecular Formula Na2O2
Molecular Weight (g/mol) 77.978
MDL Number MFCD00003497
InChI Key PFUVRDFDKPNGAV-UHFFFAOYSA-N
Synonym sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
PubChem CID 14803
IUPAC Name disodium;peroxide
SMILES [O-][O-].[Na+].[Na+]

Specifications

Melting Point 460°C (decomposition)
Color White to Yellow
Physical Form Granules
Quantity 10 g
Assay Percent Range 93% min.
Solubility Information Soluble in water,forming NaOH and H2O2. Soluble in acid. Insoluble in alkali.
Formula Weight 77.98
Grade ACS
Sensitivity Air sensitive; Hygroscopic
Appearance White to pale yellow granules
Density 2.805 g/cm3 at 20°C
Chemical Name or Material Sodium peroxide, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.