missing translation for 'onlineSavingsMsg'
Learn More

Sodium Periodate, TRC

Product Code. 30010547
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010547 100 g 100g
30006839 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010547 Supplier Toronto Research Chemicals Supplier No. TRCS665000100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-28-5
Molecular Formula I O4 . Na
Molecular Weight (g/mol) 213.8918
Synonym Periodic acid (HIO4), sodium salt (1:1), Periodic acid (HIO4), sodium salt (8CI,9CI), Sodium periodate (NaIO4) (6CI), Monosodium metaperiodate, Periodic acid sodium salt, Sodium metaperiodate, Sodium metaperiodate (NaIO4);
IUPAC Name sodium;periodate
SMILES [Na+].[O-]I(=O)(=O)=O

Specifications

Quantity 100 g
InChI Formula InChI=1S/HIO4.Na/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1
Formula Weight 213.8739
Chemical Name or Material Sodium Periodate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.