missing translation for 'onlineSavingsMsg'
Learn More

Sodium Periodate, Honeywell™ Fluka™

Product Code. 15605130 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15605130 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15605130 Supplier Honeywell-Fluka Supplier No. 36364225G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Matrix silica gel support, 10 wt. %

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 7790-28-5
Quantity 25 g
Molecular Formula INaO4
Linear Formula NaIO4
MDL Number MFCD00003534
UN Number UN3085
Synonym sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
SMILES [Na+].[O-][I](=O)(=O)=O
IUPAC Name sodium periodate
Molecular Weight (g/mol) 213.89
PubChem CID 23667635
ChEBI CHEBI:75226
Formula Weight 213.89g/mol
Packaging Glass bottle
Chemical Name or Material Sodium periodate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.