missing translation for 'onlineSavingsMsg'
Learn More

Sodium perfluorooctanoate, 97%, Thermo Scientific™

Product Code. p-7046968
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11323886

Brand: Thermo Scientific Alfa Aesar L16988.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Sodium perfluorooctanoate on gel-to-liquid-crystalline phase transition of dipalmitoylphosphatidylcholine vesicle membrane.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-95-5
Molecular Formula C8F15NaO2
Molecular Weight (g/mol) 436.052
MDL Number MFCD00040394
InChI Key LWHQXUODFPPQTL-UHFFFAOYSA-M
Synonym sodium perfluorooctanoate, sodium pentadecafluorooctanoate, sodium perfluorocaprylate, octanoic acid, pentadecafluoro-, sodium salt, perfluorooctanoic acid sodium salt, unii-63a7smf12m, pentadecafluorooctanoic acid, sodium salt, sodium pentadecafluoro octanoate, octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, sodium salt 1:1, potassium pentadecafluorooctanoate
PubChem CID 23667657
IUPAC Name sodium;2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
SMILES C(=O)(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)[O-].[Na+]

Specifications

Melting Point ∽275°C (decomposition)
Linear Formula CF3(CF2)6CO2Na
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 436.05
Percent Purity 97%
Chemical Name or Material Sodium perfluorooctanoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.