missing translation for 'onlineSavingsMsg'
Learn More

Sodium perchlorate monohydrate purum p.a., crystallized, 98.0% (T), Solstice

Product Code. 16646183
Change view
Click to view available options
50 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16646183 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 16646183 Supplier Hydranal Supplier No. 7185250KG
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

Melting Point 469°C.anhydrous substance
CAS 7-3-7791
Color Colorless
CAS Max % ≤ 100%
pH 6 to 8
Quantity 50 kg
Molecular Formula ClH2NaO5
Linear Formula NaClO4 · H2O
InChI Key IXGNPUSUVRTQGW-UHFFFAOYSA-M
SMILES O.[Na+].[O-][Cl](=O)(=O)=O
IUPAC Name sodium hydrate perchlorate
Molecular Weight (g/mol) 140.45
Grade Reagent
Density 2.02 g/cm3 (20°C)
Chemical Name or Material Sodium perchlorate monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.