missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perchlorate Monohydrate, purum p.a., ≥98% (AT), Honeywell™

Product Code. p-7072268 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15686850 250 g 250g
15676850 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15686850 Supplier Honeywell Research Chemicals Supplier No. 71852250G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Purum p.a., crystallized,≥98.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7-3-7791
Molecular Formula ClH2NaO5
Molecular Weight (g/mol) 140.45
MDL Number MFCD00149164
InChI Key IXGNPUSUVRTQGW-UHFFFAOYSA-M
Synonym sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate
PubChem CID 516899
IUPAC Name sodium;perchlorate;hydrate
SMILES O.[O-]Cl(=O)(=O)=O.[Na+]

Specifications

Quantity 250 g
Linear Formula NaClO4 · H2O
UN Number UN1502
Formula Weight 140.46g/mol
Percent Purity ≥98% (T)
Chemical Name or Material Sodium perchlorate monohydrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium perchlorate monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sodium perchlorate-1-hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.