missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium perchlorate monohydrate, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
Sodium perchlorate, monohydrate is a chaotropic agent used in standard DNA extraction and hybridization reactions in molecular biology. It is widely utilized as a deproteinization agent during nucleic acid synthesis. A bit of modern commercial explosives products may contain sodium perchlorate, ammonium perchlorate, or potassium perchlorate as a chemical sensitizing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium perchlorate, monohydrate is a chaotropic agent used in standard DNA extraction and hybridization reactions in molecular biology. It is widely utilized as a deproteinization agent during nucleic acid synthesis. A bit of modern commercial explosives products may contain sodium perchlorate, ammonium perchlorate, or potassium perchlorate as a chemical sensitizing agent.
Solubility
Soluble in water.
Notes
It is a hygroscopic solid. Store under inert atmosphere and keep away from ignition sources.
Sodium perchlorate, monohydrate is a chaotropic agent used in standard DNA extraction and hybridization reactions in molecular biology. It is widely utilized as a deproteinization agent during nucleic acid synthesis. A bit of modern commercial explosives products may contain sodium perchlorate, ammonium perchlorate, or potassium perchlorate as a chemical sensitizing agent.
Solubility
Soluble in water.
Notes
It is a hygroscopic solid. Store under inert atmosphere and keep away from ignition sources.
Specifications
Melting Point | 130°C (decomposition) |
Quantity | 1000 g |
UN Number | UN1502 |
Merck Index | 14,8653 |
Solubility Information | Soluble in water. |
Formula Weight | 140.48 (122.46 Anhydrous) |
Percent Purity | ≥97% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 2.02 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'