missing translation for 'onlineSavingsMsg'
Learn More

Sodium perchlorate, ACS Reagent, ≥98.0%, Honeywell Fluka™

Product Code. 16695863
Change view
Click to view available options
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16695863 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 16695863 Supplier Fluka Supplier No. 4102412.5KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 469°C (Decomposition)
CAS 7601-89-0
Color Colorless
Physical Form Solid
Quantity 25 kg
Molecular Formula ClNaO4
UN Number UN1502
InChI Key BAZAXWOYCMUHIX-UHFFFAOYSA-M
SMILES [Na+].[O-][Cl](=O)(=O)=O
IUPAC Name sodium perchlorate
Molecular Weight (g/mol) 122.44
Percent Purity ≥98%
Grade ACS
Chemical Name or Material Sodium perchlorate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.