missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perchlorate, 98-102%, (ACS Reagent Grade), MP Biomedicals

Product Code. 11446651 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446651 100 g 100g
11456651 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11446651 Supplier MP Biomedicals Supplier No. 0219486180
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Chemical Identifiers

CAS 7601-89-0
Molecular Formula ClNaO4
Molecular Weight (g/mol) 122.436
InChI Key BAZAXWOYCMUHIX-UHFFFAOYSA-M
Synonym sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian
PubChem CID 522606
IUPAC Name sodium;perchlorate
SMILES [O-]Cl(=O)(=O)=O.[Na+]

Specifications

Quantity 100 g
Assay Percent Range 98-102%
Formula Weight 122.4
Grade ACS Reagent
Chemical Name or Material Sodium Perchlorate, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.