missing translation for 'onlineSavingsMsg'
Learn More

Sodium percarbonate, 13-14% active oxygen

Product Code. p-7023023
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
2500 g
missing translation for 'unitSize'
100g
2500g
500g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 11433970

missing translation for 'mfr': Thermo Scientific Alfa Aesar A16045.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 15630-89-4 | Na2CO3·1.5H2O2

Sodium percarbonate is used as an oxidizing agent and environmentally friendly bleaching agent. It is used as a laboratory source of anhydrous hydrogen peroxide. It is utilized for the preparation of optically active 4-hydroxy-2-isoxazolines. It is widely used in detergent formulations and cleaning products such as carpet cleaner, textile bleach, denture cleaners, pulp and paper bleaching processes. It is also used as a hard filth removing agent in electroplating, waste water treatment agent and polymerization control agent. Further, it finds application in furniture polish and cleaners and personal care formulations. In addition to this, it is used in the preparation of trimethyl cyclopentenyl acetaldehyde and epoxydihydrolinalool.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium percarbonate is used as an oxidizing agent and environmentally friendly bleaching agent. It is used as a laboratory source of anhydrous hydrogen peroxide. It is utilized for the preparation of optically active 4-hydroxy-2-isoxazolines. It is widely used in detergent formulations and cleaning products such as carpet cleaner, textile bleach, denture cleaners, pulp and paper bleaching processes. It is also used as a hard filth removing agent in electroplating, waste water treatment agent and polymerization control agent. Further, it finds application in furniture polish and cleaners and personal care formulations. In addition to this, it is used in the preparation of trimethyl cyclopentenyl acetaldehyde and epoxydihydrolinalool.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong reducing agents, strong acids, organic materials and powdered metals.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 15630-89-4
Molecular Formula Na2CO3·1.5H2O2
MDL Number MFCD00043204
Synonym Hydrogen peroxide-Sodium carbonate adduct

Spezifikation

Quantity 100 g
UN Number UN1479
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 157.01
Chemical Name or Material Sodium percarbonate, 13 to 14% active oxygen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Sodium percarbonate, 13-14% active oxygen
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 3
Gefahrenhinweis
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P375-Fight fire remotely due to the risk of explosion.
Ergänzende Informationen
  • MIXTURE LIST-Contains : Sodium percarbonate

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt