missing translation for 'onlineSavingsMsg'
Learn More

Sodium percarbonate, 13-14% active oxygen

Product Code. 11468696
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11468696

Brand: Thermo Scientific Alfa Aesar A16045.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15630-89-4 | Na2CO3·1.5H2O2

Sodium percarbonate is used as an oxidizing agent and environmentally friendly bleaching agent. It is used as a laboratory source of anhydrous hydrogen peroxide. It is utilized for the preparation of optically active 4-hydroxy-2-isoxazolines. It is widely used in detergent formulations and cleaning products such as carpet cleaner, textile bleach, denture cleaners, pulp and paper bleaching processes. It is also used as a hard filth removing agent in electroplating, waste water treatment agent and polymerization control agent. Further, it finds application in furniture polish and cleaners and personal care formulations. In addition to this, it is used in the preparation of trimethyl cyclopentenyl acetaldehyde and epoxydihydrolinalool.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium percarbonate is used as an oxidizing agent and environmentally friendly bleaching agent. It is used as a laboratory source of anhydrous hydrogen peroxide. It is utilized for the preparation of optically active 4-hydroxy-2-isoxazolines. It is widely used in detergent formulations and cleaning products such as carpet cleaner, textile bleach, denture cleaners, pulp and paper bleaching processes. It is also used as a hard filth removing agent in electroplating, waste water treatment agent and polymerization control agent. Further, it finds application in furniture polish and cleaners and personal care formulations. In addition to this, it is used in the preparation of trimethyl cyclopentenyl acetaldehyde and epoxydihydrolinalool.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong reducing agents, strong acids, organic materials and powdered metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15630-89-4
Molecular Formula Na2CO3·1.5H2O2
MDL Number MFCD00043204
Synonym Hydrogen peroxide-Sodium carbonate adduct

Specifications

Quantity 500 g
UN Number UN1479
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 157.01
Chemical Name or Material Sodium percarbonate, 13 to 14% active oxygen
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium percarbonate, 13-14% active oxygen
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contains : Sodium percarbonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.