missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perborate Tetrahydrate, TRC

Product Code. 30424221
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424221 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30424221 Supplier Toronto Research Chemicals Supplier No. TRCS66550050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Perborate Tetrahydrate
CAS 10486-00-7
Quantity 50 g
Molecular Formula BNaO3 . 4(H2O)
Synonym Perboric Acid (HBO3) Sodium Salt Tetrahydrate, Sodium Perborate (NaBO3) Tetrahydrate, Perborn Sodium Borate (NaBO3) Tetrahydrate, Sodium Perborate Tetrahydrate, Sodium Peroxoborate Tetrahydrate
InChI Formula InChI=1S/BO3.Na.4H2O/c2-1-3-4-1;;;;;/h;;4*1H2/q-1;+1;;;;
SMILES O.O.O.O.[Na+].[O-]B1OO1
IUPAC Name sodium;3-oxidodioxaborirane;tetrahydrate
Molecular Weight (g/mol) 153.86
Formula Weight 154.0261
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.