missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium perborate tetrahydrate, Reagent Grade, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Sodium perborate tetrahydrate is mainly used in textile mills as a bleaching agent for a variety of fabrics, namely, wool, cotton, linen, rayon, etc, and as oxidizing and fixing agent for dyes. It is also used in dentifrices, denture cleaners and pharmaceuticals due to its cleansing, deodorizing and antiseptic properties and is incorporated in soaps and detergent powders.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 60°C (decomposition) |
Physical Form | Granules |
Quantity | 500 g |
Molecular Formula | NaBO3·4H2O |
Solubility Information | Soluble in water (23g/L) at 20°C. |
Formula Weight | 153.86 (81.84 Anhydrous) |
Grade | Reagent |
Sensitivity | Hygroscopic |
Density | 1.73 |
Chemical Name or Material | Sodium Perborate Tetrahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'