missing translation for 'onlineSavingsMsg'
Learn More

Sodium perborate tetrahydrate, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11367708
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Sodium perborate tetrahydrate is mainly used in textile mills as a bleaching agent for a variety of fabrics, namely, wool, cotton, linen, rayon, etc, and as oxidizing and fixing agent for dyes. It is also used in dentifrices, denture cleaners and pharmaceuticals due to its cleansing, deodorizing and antiseptic properties and is incorporated in soaps and detergent powders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10486-00-7
MDL Number MFCD00149231

Specifications

Melting Point 60°C (decomposition)
Physical Form Granules
Quantity 25 g
Molecular Formula NaBO3·4H2O
Solubility Information Soluble in water (23g/L) at 20°C.
Formula Weight 153.86 (81.84 Anhydrous)
Grade Reagent
Sensitivity Hygroscopic
Density 1.73
Chemical Name or Material Sodium Perborate Tetrahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'