missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium perborate tetrahydrate, 97%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 60.0°C |
Color | White |
pH | 10.1 to 10.6 (1% soln.) |
Physical Form | Crystalline Powder |
Quantity | 250 g |
Assay Percent Range | 97% |
Linear Formula | NaBO3·4H2O |
Fieser | 01,1102; 02,387; 04,59; 14,290; 16,310 |
Merck Index | 14,8652 |
Solubility Information | Solubility in water: 21.5g/L (18°C) |
Show More |
Safety and Handling
Product Identifier
- Sodium perborate tetrahydrate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- Reproductive toxicity Category 1B
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'