Learn More
Sodium perborate tetrahydrate, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Sodium perborate tetrahydrate is considered as a source of active hydrogen used in detergents, cleaning products, laundry detergents and bleaches. It finds application as a tooth bleaching agent utilized for tooth. It has antiseptic properties and acts as disinfectant as well as preservative in eye drops. Further, it is mixed with suitable activator, tetraacetylethylenediamine in order to release oxygen at lower temperatures.
Solubility
Soluble in water.
Notes
Moisture sensitive.Incompatible with metals, metallic salts, acids, bases and reducing agents.
Specifications
Specifications
| Melting Point | 60°C (decomposition) |
| pH | ∼10 |
| Quantity | 500 g |
| UN Number | UN1479 |
| Merck Index | 14,8652 |
| Solubility Information | Soluble in water. |
| Formula Weight | 153.86 (81.84 Anhydrous) |
| Percent Purity | 97% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Show More |
Safety and Handling
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.