missing translation for 'onlineSavingsMsg'
Learn More

Sodium perborate monohydrate, 95%

Product Code. 10337273 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10337273 1 kg Plastic bottle 1kg
10678624 25 g Plastic bottle 25g
10678814 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10337273 Supplier Thermo Scientific Acros Supplier No. 386410010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 10332-33-9 | BNaO3·H2O | 99.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10332-33-9
Molecular Formula BNaO3·H2O
Molecular Weight (g/mol) 99.81
MDL Number MFCD00149230

Specifications

Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 94% min. (active oxygen) (Iodometry)
Solubility Information Solubility in water: 15g/L (20°C)
Formula Weight 99.81
Percent Purity 95%
Packaging Plastic bottle
Chemical Name or Material Sodium perborate monohydrate
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium perborate monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 3
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.