missing translation for 'onlineSavingsMsg'
Learn More

Sodium pentacyanonitrosylferrate(III) dihydrate, ACS, 99.0-102.0%

Product Code. 11385019
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11385019 250 g 250g
11395019 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11385019 Supplier Thermo Scientific Alfa Aesar Supplier No. 087683.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Chromatographic Detection of Peptides | CAS: 13755-38-9 | C5H4FeN6Na2O3 | 297.95 g/mol

For chromatographic detection of peptides.Sodium pentacyanonitrosylferrate(III) dihydrate, is used as a vasodilator drug to reduce blood pressure. It is used as an analytical reagent for the detection of methyl ketones and amines. Further, it is used as a reference compound for the calibration of Mossbauer spectrometer. It is also used for chromatographic detection of peptides containing cystine. It finds application as a color reagent in the development of thin layer chromatography in combination with acetaldehyde. It is also used in presence of buffers as reagent for ketone strips, which is utilized to test ketone in the urine of a diabetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13755-38-9
Molecular Formula C5H4FeN6Na2O3
Molecular Weight (g/mol) 297.95
MDL Number MFCD00149192
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
PubChem CID 11953895
IUPAC Name disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

Specifications

Color Red
Physical Form Crystals
Quantity 250 g
Assay Percent Range 99.0-102.0%
Solubility Information Soluble in water. Slightly soluble ethanol.
Formula Weight 297.95 (261.92 Anhydrous)
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Density 1.72
Chemical Name or Material Sodium pentacyanonitrosylferrate(III) dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.