missing translation for 'onlineSavingsMsg'
Learn More

Sodium (para)periodate, ≥99.0% (as H2INa3O6, RT), Honeywell™

Product Code. p-7120765 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 15606860

Brand: Honeywell Research Chemicals 71872100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥99.0% (as H2INa3O6, RT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13940-38-0
Molecular Formula H2INa3O6
Molecular Weight (g/mol) 293.88
MDL Number MFCD00036146
InChI Key DSYMXPNUZLZKOA-UHFFFAOYSA-K
Synonym sodium paraperiodate, sodium para periodate, sodium triparaperiodate, sodium hydrogen periodate, 3na.h2io6, periodic acid trisodium salt, trisodium dihydrogen orthoperiodate, sodium para periodate, commercial, trisodium dihydroxydioxidoiodosylolate, trisodium dihydroxy-trioxido-oxo-$l^ 7-iodane
PubChem CID 9796123
IUPAC Name trisodium (dihydroxydioxidoiodosyl)olate
SMILES [Na+].[Na+].[Na+].O[I](O)([O-])([O-])([O-])=O

Specifications

Melting Point 300°C
Color White
Physical Form Crystals
Quantity 100 g
Linear Formula Na3H2IO6
UN Number UN1479
Solubility Information 106.7g/L at 20°C
Formula Weight 293.89g/mol
Percent Purity ≥99.0% (as H2INa3O6, RT)
Packaging Poly bottle
Chemical Name or Material Sodium (para)periodate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.