missing translation for 'onlineSavingsMsg'
Learn More

Sodium (para)periodate, ≥99.0% (as H2INa3O6, RT), Honeywell™

Product Code. 15606860 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15606860 100 g Poly bottle 100g
15616860 500 g Poly bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15606860 Supplier Honeywell Research Chemicals Supplier No. 71872100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥99.0% (as H2INa3O6, RT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13940-38-0
Molecular Formula H2INa3O6
Molecular Weight (g/mol) 293.88
MDL Number MFCD00036146
InChI Key DSYMXPNUZLZKOA-UHFFFAOYSA-K
Synonym sodium paraperiodate, sodium para periodate, sodium triparaperiodate, sodium hydrogen periodate, 3na.h2io6, periodic acid trisodium salt, trisodium dihydrogen orthoperiodate, sodium para periodate, commercial, trisodium dihydroxydioxidoiodosylolate, trisodium dihydroxy-trioxido-oxo-$l^ 7-iodane
PubChem CID 9796123
IUPAC Name trisodium (dihydroxydioxidoiodosyl)olate
SMILES [Na+].[Na+].[Na+].O[I](O)([O-])([O-])([O-])=O

Specifications

Melting Point 300°C
Color White
Physical Form Crystals
Quantity 100 g
Linear Formula Na3H2IO6
UN Number UN1479
Solubility Information 106.7g/L at 20°C
Formula Weight 293.89g/mol
Percent Purity ≥99.0% (as H2INa3O6, RT)
Packaging Poly bottle
Chemical Name or Material Sodium (para)periodate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.