missing translation for 'onlineSavingsMsg'
Learn More

Sodium p-Toluenesulfonate, TRC

Product Code. 30415230
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30415230 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415230 Supplier Toronto Research Chemicals Supplier No. TRCS2245805G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium p-Toluenesulfonate
CAS 657-84-1
Molecular Formula C7 H7 O3 S . Na
Quantity 5 g
Synonym Sodium p-Toluensulfonate, Sodium p-Tolyl Sulfonate, Sodium Toluenesulfonate, Sodium Tosylate, Taycatox N 3000, Toluenesulfonic Acid Sodium Salt, p-Toluenesulfonic Acid Sodium Salt (7CI,8CI), 4-Methylbenzenesulfonic Acid Sodium Salt, 4-Toluenesulfonic Acid Sodium Salt, Chemguard NATS, Cyclophil STS 70, Eltesol ST 34, Eltesol ST 90, Naxonate ST, Pilot STS 32, Sodium 4-Methylbenzenesulfonate, Sodium 4-Toluenesulfonate, Sodium Methylbenzenesulfonate, Sodium p-Methylbenzenesulfonate
InChI Formula InChI=1S/C7H8O3S.Na/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);/q;+1/p-1
SMILES [Na+].Cc1ccc(cc1)S(=O)(=O)[O-]
IUPAC Name sodium;4-methylbenzenesulfonate
Molecular Weight (g/mol) 194.183
Formula Weight 194.001
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.