missing translation for 'onlineSavingsMsg'
Learn More

Sodium oxide

Product Code. 11314247
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11314247

Brand: Thermo Scientific Alfa Aesar 036712.22

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1313-59-3 | Na2O | 61.979 g/mol

Sodium oxide is used in chemical manufacturing, ceramics and glasses. Ethylbenzene dehydrogenation and carbon dioxide shift-reaction occurs using a sodium oxide/alumina catalyst. It is employed as catalyzing gasification of CO2 on carbon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1313-59-3
Molecular Formula Na2O
Molecular Weight (g/mol) 61.979
MDL Number MFCD00046201
InChI Key KKCBUQHMOMHUOY-UHFFFAOYSA-N
Synonym sodium oxide, disodium oxide, disodium monoxide, disodium oxygen-2 anion, disodium oxidandiide, sodium oxide 25g, disodium oxygen 2-, 2na.o
PubChem CID 73971
IUPAC Name disodium;oxygen(2-)
SMILES [O-2].[Na+].[Na+]

Specifications

Melting Point >400°C (decomposition)
Color White to Gray
Physical Form Beads
Quantity 100 g
UN Number UN1825
Merck Index 14,8651
Solubility Information Soluble in water.
Formula Weight 61.98
Odor Odorless
Sensitivity Air and moisture sensitive
Density 2.27 g/mL
Chemical Name or Material Sodium oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.