missing translation for 'onlineSavingsMsg'
Learn More

Sodium Oxalate (White Crystals or Powder), Fisher BioReagents

Product Code. 10583505
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10583505 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10583505 Supplier Fisher BioReagents Supplier No. BP353500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
This sodium salt is used in buffers for molecular biology and cell culture applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 62-76-0
Melting Point 250°C
Color White
pH 8
Loss on Drying 0.05% max.
Assay Percent Range ≥99.0 %
Packaging Glass Bottle
Molecular Formula C2Na2O4
MDL Number MFCD00012465
Quantity 500 g
Merck Index 15, 8781
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
Solubility Information 0.005% max. Insoluble Matter
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O
IUPAC Name disodium;oxalate
Molecular Weight (g/mol) 134.00
PubChem CID 6125
Formula Weight 134
Percent Purity ≥99.0%
Physical Form Powder or Solid
Chemical Name or Material Sodium Oxalate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.