missing translation for 'onlineSavingsMsg'
Learn More

Sodium Oxalate, puriss p.a, ≥99.5% (RT), Honeywell Fluka™

Product Code. p-7120817 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
Unit Size:
100g
500g
Product Code. Quantity Packaging unitSize
15636850 100 g Poly bottle 100g
15646850 500 g Poly bottle 500g
2 options
This item is not returnable. View return policy

Product Code. 15636850

Brand: Honeywell-Fluka 71800100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.5% (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium;oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Boiling Point Decomposes on heating
Melting Point 308°C
Color Colorless
pH 5.0 to 7.0 (at 20°C)
Physical Form Solid
Quantity 100 g
Linear Formula NaOOCCOONa
UN Number NONH for all modes of transport
Beilstein 3631622
Solubility Information 883g/L (at 20°C)
Formula Weight 134g/mol
Percent Purity ≥99.5% (RT)
Odor Odorless
Packaging Poly bottle
Density 2.26g/cm3
Chemical Name or Material Sodium oxalate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.