missing translation for 'onlineSavingsMsg'
Learn More

Sodium Oxalate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15518174 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
Unit Size:
125g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15518174 125 g Wide Mouth Amber Glass Bottle 125g
15508174 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15518174 Supplier Avantor J.T.Baker Supplier No. 380104

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Physical Form Powder
Quantity 125 g
IUPAC Name disodium oxalate
Formula Weight 134
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.34kg
Chemical Name or Material Sodium Oxalate, Titrimetry Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.