missing translation for 'onlineSavingsMsg'
Learn More

Sodium Oxalate, Honeywell™ Fluka™

Product Code. 15607990 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15607990 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15607990 Supplier Honeywell-Fluka Supplier No. O0136100G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BioXtra

  • Stringent testing protocols may include trace metal analysis and specialized suitability testing, such as hybridoma or mouse embryo
  • Includes application-specific formulations, such as tablets and prepared solutions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 62-76-0
Quantity 100 g
Molecular Formula C2Na2O4
Linear Formula NaOOCCOONa
MDL Number MFCD00012465
UN Number NONH for all modes of transport
Beilstein 3631622
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O
IUPAC Name disodium;oxalate
Molecular Weight (g/mol) 134.00
PubChem CID 6125
Formula Weight 134g/mol
Packaging Glass bottle
Chemical Name or Material Sodium oxalate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.