missing translation for 'onlineSavingsMsg'
Learn More

Sodium orthovanadate, MedChemExpress

Product Code. 16491188
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491188 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16491188 Supplier Medchem Express Supplier No. HYD0852
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sodium orthovanadate is an inhibitor of protein tyrosine phosphatases, alkaline phosphatases and a number of ATPases, most likely acting as a phosphate analogue.

Sodium orthovanadate is an inhibitor of protein tyrosine phosphatases, alkaline phosphatases and a number of ATPases, most likely acting as a phosphate analogue.

Specifications

Chemical Name or Material Sodium orthovanadate
CAS 13721-39-6
Color Off-White
Purity Grade Notes Research
Molecular Formula Na3O4V
Quantity 5 g
Solubility Information H2O : 8.33 mg/mL (45.29 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 80°C) (insoluble or slightly soluble)
SMILES [O].[Na].[V]
Molecular Weight (g/mol) 183.91
Formula Weight 183.91
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.