missing translation for 'onlineSavingsMsg'
Learn More

Sodium Orthovanadate, 96% min., (Powder), MP Biomedicals

Product Code. 11495721 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g

Product Code. 11495721

Brand: MP Biomedicals 0215966480

Please to purchase this item. Need a web account? Register with us today!

Potent alkaline phosphate inhibitor

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13721-39-6
Molecular Formula Na3O4V
Molecular Weight (g/mol) 183.907
InChI Key IHIXIJGXTJIKRB-UHFFFAOYSA-N
Synonym sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt
PubChem CID 61671
ChEBI CHEBI:35607
IUPAC Name trisodium;trioxido(oxo)vanadium
SMILES [O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Physical Form Powder
Quantity 100 g
Assay Percent Range 96% min.
Formula Weight 183.9
Chemical Name or Material Sodium Orthovanadate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.