missing translation for 'onlineSavingsMsg'
Learn More

Sodium orthosilicate, (mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution), Thermo Scientific Chemicals

Product Code. 11373377
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373377 5 kg 5kg
11363377 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373377 Supplier Thermo Scientific Alfa Aesar Supplier No. 017568.A7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is used as a degreasant, industrial alkaline detergents, water treatment (boiler-feed); adhesives, builder in soaps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-30-5
Molecular Formula Na4O4Si
Molecular Weight (g/mol) 184.04
MDL Number MFCD00014252
InChI Key POWFTOSLLWLEBN-UHFFFAOYSA-N
Synonym sodium orthosilicate, tetrasodium orthosilicate, tetrasodium silicate, silicic acid h4sio4 , sodium salt 1:4, silicic acid h4sio4 , tetrasodium salt, sodium silicate, ortho, teu2jic5pa, unii-teu2jic5pa, dryorth, sodium silicate 4-
PubChem CID 26051
IUPAC Name tetrasodium;silicate
SMILES [O-][Si]([O-])([O-])[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Color White
Physical Form Crystalline
Quantity 5 kg
UN Number UN1759
Odor Odorless
Chemical Name or Material Sodium orthosilicate, Mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.