missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitrite, TRC
Click to view available options
Quantity:
500 g
Unit Size:
500g
Specifications
Specifications
| Chemical Name or Material | Sodium nitrite |
| CAS | 7632-00-0 |
| Molecular Formula | N O2 . Na |
| Quantity | 500 g |
| Synonym | Sodium nitrite, Nitrous Acid Sodium Salt, Anti-Rust, E 250, Erinitrit, Filmerine, M 138C, Nitrous Acid Soda, SNS 40, Sodium Nitrite, Sodium Nitrite (NaNO2), Synfat 1004 |
| InChI Formula | InChI=1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1 |
| SMILES | [Na+].[O-]N=O |
| IUPAC Name | sodium;nitrite |
| Molecular Weight (g/mol) | 69 |
| Formula Weight | 68.9827 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction