missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitrite Solution, Volumetric, 0.2M NaNO2 (0.2N), Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Unit Size:
1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| CAS | 7632-00-0 |
| Quantity | 1 L |
| Molecular Formula | NNaO2 |
| Linear Formula | NaNO2 |
| MDL Number | MFCD00011118 |
| UN Number | NONH for all modes of transport |
| Synonym | sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts |
| InChI Key | LPXPTNMVRIOKMN-UHFFFAOYSA-M |
| SMILES | [Na+].[O-]N=O |
| IUPAC Name | sodium nitrite |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction