missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, Extra Pure, SLR, Fisher Chemical™

Product Code. 10182920 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10182920 2.5 kg 2.5kg
10607042 500 g 500g
10305770 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10182920 Supplier Fisher Chemical Supplier No. S/5600/62
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium;nitrite
SMILES [Na+].[O-]N=O

Specifications

Boiling Point 320°C
Melting Point 271°C
Color Yellow
pH 8 to 9
Physical Form Solid
Quantity 2.5 kg
Formula Weight 69g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium nitrite
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 3
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H301-Toxic if swallowed.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P273-Avoid release to the environment.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.