missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, Certified AR for Analysis, Fisher Chemical™

Product Code. 10498990 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10498990 500 g 500g
10274110 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Product Code. 10498990 Supplier Fisher Chemical Supplier No. S/5640/53
  • ONLINE EXCLUSIVE PRICE
    £74.20
    £59.40 / 500g
    Save £14.80
    19% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

CAS: 7632-00-0 | Molecular Formula: NNaO2 | Molecular Weight: 69.00 g/mol

Sodium Nitrite, Certified AR for Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium;nitrite
SMILES [Na+].[O-]N=O

Specifications

Boiling Point 320°C
Melting Point 271°C
Color Yellow
pH 8 to 9
Physical Form Solid
Quantity 500 g
Packaging Plastic powder jar
Formula Weight 69g/mol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium nitrite
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 3
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H301-Toxic if swallowed.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.